missing translation for 'onlineSavingsMsg'
Learn More

Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl

Product Code. 10072600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10072600 500 mL 500mL
10695832 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072600 Supplier Thermo Scientific Acros Supplier No. 196285000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-49-2 | Se | 78.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Selenium standard solution
Name Note 1 mg/ml Se in 10% HCl
Packaging Plastic bottle
CAS 7782-49-2, 7647-01-0, 7732-18-5
Density 4.81
Physical Form Solution
Molecular Formula Se
Concentration 995 to 1005 μg/mL of Se in 10% HCl (20°C)
MDL Number MFCD00134090 MFCD00011224
Merck Index 14, 8431
Solubility Soluble in water
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES [Se]
IUPAC Name selenium
Molecular Weight (g/mol) 78.97
PubChem CID 6326970
Quantity 500 mL
ChEBI CHEBI:27568
Formula Weight 78.96
Grade AAS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Selenium standard solution, 1 mg/ml Se in 10% HCl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.