missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Screw Caps for Screwthread Tubes

Product Code. 10192041
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10192041 Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192041 Supplier Lenz Laborglasintrumente™ Supplier No. 01320025
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Screw caps with DIN thread (GL)

  • Temperature resistant up to 180 °C
  • With PTFE coated silicone sealing (VMQ)
  • Material: PBT
  • Color: Red

Specifications

Closure Size GL25
Product Type Screw Cap
Material PBT
Color Red
Quantity 10
For Use With (Equipment) Screw Thread Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.