Learn More
BOLA Screw Caps
Product Code. 10466591
Change view
Click to view available options
Thread Size (Metric):
GL 14
GL 18
GL 25
GL 32
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
10466591 GL 14 Each
10105601 GL 18 Each
10083521 GL 25 Each
10727532 GL 32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10466591 Supplier BOLA Supplier No. H98601
Only null left
Add to basket
Add to basket
Screw Caps PTFE
Screw cap with handy knurl, suitable for GL threads, made of PTFE. After assembly, the product is only exposed to PTFE.
Specifications
Color White
Sterility Non-sterile
Thread Size (Metric) GL 14
Neck Finish 98 mm
Certifications/Compliance FDA
Disposable Reusable
Autoclavable Autoclavable