Learn More
Lenz Laborglasintrumente Screw Caps
Description
- Material: PBT (red)
- With PTFE-coated sealing
- Temperature resistance: up to 180 °C
Specifications
Specifications
| Color | Red |
| Material | PBT |
| Thread Size (Metric) | 32 GL |
| Closure Material | PTFE |