Learn More
Kartell™ PE Standardized Screw Cap
Product Code. 10172471
Change view
Click to view available options
Thread Size (Metric):
GL 25
Unit Size:
Pack of 50
Product Code. Thread Size (Metric) unitSize
10172471 GL 25 Pack of 50
1 options
Product Code. 10172471 Supplier Kartell™ Supplier No. 0330100
Only null left
Add to basket
Add to basket
Constructed of polyethylene, making it be lightweight and strong. Kartell™ PE Standardized Screw Cap is used to tightly seal a bottle, reducing the risk of spillage and sample contamination.
  • Ligtweight
  • For use with wash bottles
  • For Kartell™ Bottles 15801. 15821, 15841, 15861, 15881, 15901
Specifications
Material Polyethylene
Product Line Cap
Thread Size (Metric) GL 25
Neck Finish 30 mm
Product Type Wash Bottle Closure
For Use With (Equipment) Kartell™ Bottles 15801, 15821, 15841, 15861, 15881, 15901