missing translation for 'onlineSavingsMsg'
Learn More

Scandium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Product Code. 10050562 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10050562 500 mL Triple-leached LDPE bottle 500mL
10080392 125 mL Triple-leached LDPE bottle 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10050562 Supplier SPEX Certiprep Supplier No. PLSC22X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 12060-08-1 , 7697-37-2 , 7732-18-5
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS Min % 0.1, 2.0, 97.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 2.0, 97.9
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 500 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.2-5% HNO3    1000μg/mL    PLSC2-2Y, -2X    10,000    PLSC2-3Y, -3X
Chemical Name or Material Scandium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000 µg/mL Scandium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.