Learn More
Scandium(III) oxide, REacton™, 99.99% (REO)
Description
Scandium oxide is resistant to heat and thermal shock, hence finds use in high-temperature systems. Further, it is used in electronic ceramics and glass composition. It is used as a starting material for the preparation of other scandium compounds. In addition to this, it has vacuum deposition applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2485°C |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.99% |
| Solubility Information | Insoluble in water. |
| Formula Weight | 137.91 |
| Odor | Odorless |
| Refractive Index | 1.964 |
| Chemical Name or Material | Scandium(III) oxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.