missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Sampling Tube Adapter

Product Code. 10113152
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10113152 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10113152 Supplier Buerkle™ Supplier No. 53051020
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Buerkle™ PTFE Sampling Tube Adapter is a spare part for Buerkle™ MiniSampler.

  • Spare part for Buerkle™ MiniSampler
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adapter for Sample Tubes
Material PTFE
Description Equipment for MiniSampler PTFE
Color White
For Use With (Application) MiniSampler PTFE Sample Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.