missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic Acid, Extra Pure, SLR, Fisher Chemical™
49.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
49.01 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
49.01 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
Specifications
Melting Point | 158°C |
Color | White |
pH | 3.6 |
Boiling Point | 211°C |
Packaging | Plastic powder jar |
Quantity | 500 g |
Formula Weight | 138.12g/mol |
Vapor Pressure | 0.3mbar 95 |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'