missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl-L-cysteine, TRC

Product Code. 30465784
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465784 5 g 5g
30475770 1 g 1g
30472771 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30465784 Supplier Toronto Research Chemicals Supplier No. TRCM2946555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187-84-4
Molecular Formula C4 H9 N O2 S
Molecular Weight (g/mol) 135.19
Synonym S-Methylcysteine, L-Methylcysteine, H-Cys(Me)-OH, (R)-2-amino-3-(methylthio)propanoic acid, 3-(Methylthio)-L-alanine, (R)-2-Amino-3-(methylmercapto)propionic acid, (-)-S-Methyl-L-cysteine
IUPAC Name (2R)-2-amino-3-methylsulfanyl-propanoic acid
SMILES CSC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C4H9NO2S/c1-8-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1
Quantity 5 g
Formula Weight 135.0354
Shipping Condition Room Temperature
Chemical Name or Material S-Methyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.