missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RYK Polyclonal Antibody
GREENER_CHOICE

Product Code. 10160710
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10160710 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10160710 Supplier Invitrogen™ Supplier No. PA514734

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

RYK is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. A nine nucleotide insertion in some transcripts results in the SLG variant. It is not established whether this is a product of alternative splicing or a second gene, since evidence for a second gene or pseudogene on chromosome 17 exists.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RYK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RYK
Gene Accession No. P34925
Gene Alias AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik
Gene Symbols RYK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 561-591 amino acids from the C-terminal region of human RYK.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.