Learn More
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Description
Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red to Brown |
| Linear Formula | Ru(NO)(NO3)3 |
| UN Number | UN3093 |
| Concentration | Ru 1.5% w/v |
| Density | 1.07 g/mL |
| Solubility Information | Miscible with cold water. |
| Formula Weight | 317.09 |
| Odor | Acidic |
| Quantity | 1000 mL |
| Physical Form | Liquid |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.