missing translation for 'onlineSavingsMsg'
Learn More

Restek Rtx-200 Cap. Column 30m, 0.25mm ID, 1.00um

Brand:  Restek 15053

Encompass_Preferred

Rtx-200 Cap. Column 30m, 0.25mm ID, 1.00um

Product Code. 10000152

  • £769.56 / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.