missing translation for 'onlineSavingsMsg'
Learn More

(RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one, Mikromol™

Product Code. 30051795
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30051795 25 mg 25mg
17953086 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 30051795

Brand: Mikromol MM0228.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one
CAS 123155-82-8
Analyte or Component Names (RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one
Shipping Condition Room Temperature
Molecular Formula C17 H20 F N3 O2
Certifications/Compliance ISO 17025
Synonym (RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one, Ofloxacin Imp. B (EP), Ofloxacin Impurity B
InChI Formula InChI=1S/C17H20FN3O2/c1-11-10-23-17-15-12(14(22)3-4-21(11)15)9-13(18)16(17)20-7-5-19(2)6-8-20/h3-4,9,11H,5-8,10H2,1-2H3
SMILES CC1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C=CN1c24
IUPAC Name (RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one
Molecular Weight (g/mol) 317.36
Quantity 25 mg
Formula Weight 317.154 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.