missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROC1 Polyclonal Antibody

Código de producto. 10047522
Change view
Click to view available options
Quantity:
100 μg
Tamaño de la unidad:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
10047522 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
For Research Use Only. All usage must comply with product instructions.
 
Código de producto. 10047522 Proveedor Invitrogen™ N.º de proveedor 342500

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

ROC1 Polyclonal Antibody for Western Blot, IP

This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen ROC1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene RBX1
Gene Accession No. P62877, P62878
Gene Alias 1500002P15Rik; AA517855; BA554C12.1; E3 ubiquitin-protein ligase RBX1; E3 ubiquitin-protein ligase RBX1, N-terminally processed; E3 ubiquitin-protein transferase RBX1; E3 ubiquitin-protein transferase RBX1, N-terminally processed; Protein ZYP; Rbx1; Regulator of cullins 1; RING box protein 1; RING finger protein 75; ring-box 1; ring-box 1, E3 ubiquitin protein ligase; RING-box protein 1; RNF75; ROC1; ZYP protein
Gene Symbols RBX1
Host Species Rabbit
Immunogen specific to Human ROC1C.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56438, 9978
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.