Learn More
Hamilton™ Robotics Low Bleed High Temperature Septa
Product Code. 10034202
Change view
Click to view available options
Diameter (Metric):
9.5 mm
12.7 mm
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10034202 9.5 mm Pack of 50
10710844 12.7 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10034202 Supplier Hamilton™ Supplier No. 75833
Only null left
Add to basket
Add to basket
Specifications
Diameter (Metric) 9.5 mm
For Use With (Equipment) GC
Quantity 50/Pk.
Product Type Low Bleed High Temperature Septa