missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RNB 61

Product Code. 30281235
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30281235 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281235 Supplier Tocris Bioscience™ Supplier No. 8835/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent and selective CB2R agonist

RNB 61 is a highly potent and selective cannabinoid CB2 receptor (CB2R) agonist (Ki = 0.57 nM). RNB 61 shows >5000-fold selectivity for hCB2R over hCB1R. In vivo, RNB 61 inhibits the FSK-induced cAMP formation on mouse, rat, and dog CB2Rs (EC50 values ranging from 0.13 to 1.86 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(3E)-5-tert-Butyl-2-(cyclobutylmethyl)-1-methyl-1,2-dihydro-3H-pyrazol-3-ylidene]-2-(2-hydroxy-2-methylpropoxy)-5-(trifluoromethyl)benzamide
CAS 1217403-51-4
Quantity 10 mg
Target Cannabinoid R2/CB2 Agonists
Molecular Formula C25H34F3N3O3
Purity 0.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.