missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RIPK2, His Tag Recombinant Protein
RIPK2 is a serine/threonine kinase.
Brand: Invitrogen™ PV4325
Code : NEW
Additional Details : Weight : 0.00900kg
Description
RIPK2 is involved in cell activation, proliferation, differentiation, and apoptosis.
Specifications
NP_003812 | |
1mg | |
RIPK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
38.9 kDa | |
RIPK2, RICK, RIP2 | |
Human | |
His Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |