Description
- Advanced defenses: Developed with F3 Technology™, RINGERS™ R068 impact gloves feature a comfortable, dexterous TPR (thermoplastic rubber) design, offering optimal hand coverage
- Specialized protection: These hand protection gloves boast fully dipped nitrile coating, granting wearers an effective liquid resistance barrier, for truly reliable personal protection
- Enhanced features: Their high-visibility design reinforces wearer protection, while touchscreen-compatible thumb tips, index and middle fingers enable technological interaction without compromising hand safety
- Strengthened grip: RINGERS™ R068 safety gloves also feature half-dipped palm coating with a sandy finish, offering better grip on wet and dry surfaces, for improved workplace safety
Specifications
Specifications
| Coating Material | Nitrile |
| Color | Red |
| Cut Resistance Rating | Level F |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | HPPE, Nitrile |
| Product Type | Industrial Gloves |
| Size | 9 |