missing translation for 'onlineSavingsMsg'
Learn More

Riluzole, MedChemExpress

Product Code. p-200100957
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472089 1 g 1g
16444307 50 mg 50mg
16428176 100 mg 100mg
16402066 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472089 Supplier Medchem Express Supplier No. HYB0211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Riluzole is an anticonvulsant drug and belongs to the family of use-dependent Na+ channel blocker which can also inhibit GABA uptake with an IC50 of 43 μM.

Riluzole is an anticonvulsant drug and belongs to the family of use-dependent Na+ channel blocker which can also inhibit GABA uptake with an IC50 of 43 μM.

Specifications

Chemical Name or Material Riluzole
CAS 1744-22-5
Color White
Purity Grade Notes Research
Molecular Formula C8H5F3N2OS
Quantity 1 g
Synonym PK 26124
Solubility Information DMSO : 100 mg/mL (426.99 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.27 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES NC1=NC2=CC=C(OC(F)(F)F)C=C2S1
Molecular Weight (g/mol) 234.2
Formula Weight 234.2
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.