missing translation for 'onlineSavingsMsg'
Learn More

Reversan, TRC

Product Code. 30427554
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427554 5 mg
30427811 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427554 Supplier Toronto Research Chemicals Supplier No. TRCR2798005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313397-13-6
Molecular Formula C26H27N5O2
Molecular Weight (g/mol) 441.52
Synonym N-[3-(4-Morpholinyl)propyl]-5,7-diphenylpyrazolo[1,5-a]pyrimidine-3-carboxamide
IUPAC Name N-(3-morpholin-4-ylpropyl)-5,7-diphenylpyrazolo[1,5-a]pyrimidine-3-carboxamide
SMILES O=C(NCCCN1CCOCC1)c2cnn3c(cc(nc23)c4ccccc4)c5ccccc5

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H27N5O2/c32-26(27-12-7-13-30-14-16-33-17-15-30)22-19-28-31-24(21-10-5-2-6-11-21)18-23(29-25(22)31)20-8-3-1-4-9-20/h1-6,8-11,18-19H,7,12-17H2,(H,27,32)
Formula Weight 441.2165
Chemical Name or Material Reversan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.