missing translation for 'onlineSavingsMsg'
Learn More

Reversan, MedChemExpress

Product Code. 16476847
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476847 5 mg 5mg
16460126 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476847 Supplier Medchem Express Supplier No. HY107643

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Reversan (CBLC4H10) is a potent and nontoxic multidrug resistance-associated protein 1 (MRP1) and P-glycoprotein (Pgp) inhibitor.

Reversan (CBLC4H10) is a potent and nontoxic multidrug resistance-associated protein 1 (MRP1) and P-glycoprotein (Pgp) inhibitor.

Specifications

Chemical Name or Material Reversan
CAS 313397-13-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H27N5O2
Quantity 5 mg
Synonym CBLC4H10
Solubility Information DMSO : 15.62 mg/mL (35.38 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=C2N(C(C3=CC=CC=C3)=CC(C4=CC=CC=C4)=N2)N=C1)NCCCN5CCOCC5
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.