Learn More
Restek™ Valco Connectors
Product Code. 10093366
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Inner:
0.15 mm
0.25 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10093366 0.15 mm Each
13081580 0.15 mm -
13091580 0.25 mm -
10606015 0.25 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10093366 Supplier Restek™ Supplier No. 20147
Only null left
Add to basket
Add to basket
For use with 0.0625 in. OD tubing.
  • Ends of tubing seat squarely at bottoms of fitting details
  • Made of 300-series stainless steel
Specifications
Type Union
Diameter (Metric) Inner 0.15 mm
For Use With HPLC