missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Straight Inlet Liners for Splitless Injection

Product Code. 13164592
Encompass_Preferred
Change view
Click to view available options
Quantity:
25/Pk.
5/Pk.
Each
Diameter (Metric) Inner:
0.2 cm
0.4 cm
4.0 mm
Includes:
Glass Splitless
Glass Splitless with Wool
Quartz Splitless with Wool
Splitless/HP w/Glass Wool
Unit Size:
Pack of 5
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Diameter (Metric) Inner Includes unitSize
13164592 5/Pk. 0.4 cm Glass Splitless with Wool Pack of 5
12912649 Each 0.2 cm Glass Splitless -
12952739 Each 0.4 cm Glass Splitless -
13134592 Each 0.4 cm Glass Splitless with Wool -
10058314 5/Pk. 0.2 cm Glass Splitless Pack of 5
12972649 5/Pk. 0.2 cm Glass Splitless -
12932659 25/Pk. 0.2 cm Glass Splitless -
12922749 5/Pk. 0.4 cm Glass Splitless -
10097781 5/Pk. 0.4 cm Glass Splitless Pack of 5
12932749 5/Pk. 4.0 mm Splitless/HP w/Glass Wool -
12912759 25/Pk. 0.4 cm Glass Splitless -
13114602 25/Pk. 0.4 cm Glass Splitless with Wool -
13184602 5/Pk. 0.4 cm Quartz Splitless with Wool -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 13164592 Supplier Restek™ Supplier No. 22401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition

  • Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
  • Compatible with Agilent™/Finnigan™ columns
  • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples; also, packing in wool is recommended to aid in vaporization of high molecular weight compounds and to minimize discrimination
  • O.D. × Length: 6.5 × 78.5mm

Specifications

Includes Glass Splitless with Wool
Quantity 5/Pk.
Length (Metric) 7.85 cm
Packing Material Deactivated Wool
Product Type IP Splitless Straight Inlet Liner
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.65 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.