missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner

Product Code. 10145827
Encompass_Preferred
Change view
Click to view available options
Packing Material:
Base Deactivated
Base Deactivated Wool
CarboFrit
Deactivated Wool
None
Product Type:
Cup Splitter Inlet Liner
IP Cup Splitter Inlet Liner
Quantity:
1/Pk.
5/Pk.
Unit Size:
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packing Material Product Type Quantity unitSize
10145827 None IP Cup Splitter Inlet Liner 5/Pk. Pack of 5
12932629 Deactivated Wool Cup Splitter Inlet Liner 1/Pk. -
12922629 None Cup Splitter Inlet Liner 5/Pk. -
12912639 None IP Cup Splitter Inlet Liner 5/Pk. -
12922639 Deactivated Wool IP Cup Splitter Inlet Liner 5/Pk. -
12942639 CarboFrit IP Cup Splitter Inlet Liner 5/Pk. Pack of 5
12952639 Base Deactivated Cup Splitter Inlet Liner 5/Pk. Pack of 5
12962639 Base Deactivated Wool Cup Splitter Inlet Liner 5/Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10145827 Supplier Restek™ Supplier No. 20710
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For high or low molecular weight compounds

  • Increased sample residence time in the liner for homogeneous vaporization
  • 4mm I.D.
  • Available with or without packing

Specifications

Quantity 5/Pk.
Length (Metric) 7.85 cm
Packing Material None
Product Type IP Cup Splitter Inlet Liner
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.63 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.