missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Shortix Capillary GC Column Cutter

Consistently make precise, clean and square cuts with a diamond blade

Brand:  Restek™ 23026

Encompass_Preferred

 View more versions of this product

Product Code. 10152832

  • £648.56 / Each
Estimated Shipment: 21-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Built-in magnifier to verify square cut
  • Use with 0.25 mm ID to 0.53 mm ID tubing (0.78 mm OD maximum)

Specifications

Specifications

Capillary GC Column Cutter
Built in magnifier
Gas Chromatography
0.25 to 0.53mm I.D. tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers