Learn More
Restek™ MXT-Union Connector Kits For Fused Silica Columns
Description
- Low dead volume, leak-tight connection
- Reusable
- Siltek treatment ensures maximum inertness
- Ideal for connecting a guard column or transfer line to an analytical column
- Use to oven temperatures of 350°C
- Can also be used for connecting fused silica to metal
- Includes MXT union, two 1/32 in. nuts and two one-piece, fused silica adapters
Specifications
Specifications
| Type | Connector |
| Includes | MXT union, two 1/32 in. nuts and two one-piece, fused silica adapters |
| For Use With | 0.32 mm I.D. Fused Silica Columns |