missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Inlet Liner Seals for Varian 1177 Injectors

Product Code. 13071700
Encompass_Preferred
Change view
Click to view available options
Quantity:
10/Pk.
50/Pk.
Diameter (Metric) Inner:
0.635 cm
0.65 cm
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Diameter (Metric) Inner unitSize
13071700 50/Pk. 0.635 cm -
10649542 50/Pk. 0.635 cm -
13081700 10/Pk. 0.65 cm -
10045981 50/Pk. 0.65 cm Pack of 50
13091700 50/Pk. 0.65 cm -
13061700 10/Pk. 0.635 cm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 13071700 Supplier Restek™ Supplier No. 20297

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Agilent and Bruker/Varian 1177 Injectors.

  • High thermal stability at injection port temperatures up to 450°C
  • For split or splitless liners

Specifications

Quantity 50/Pk.
Temperature (Metric) Max 450°C
Product Type O-ring
Diameter (Metric) Inner 0.635 cm
For Use With (Equipment) Split Liners, Agilent and Bruker/Varian 1177 Injectors
Liner Material Graphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.