Learn More
Restek™ Air Diverters for Agilent 5890/6890/6850 GCs
Product Code. 10012082
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Product Code. Packaging Quantity unitSize
10012082 1 Each Each
1 options
Product Code. 10012082 Supplier Restek™ Supplier No. 22076
For use with Agilent 5890, 6890, 7890 GCs.
  • Divert GC exhaust heat up and away from the lab bench
  • Easy to install; no tools required
  • Compatible with 4 in. flexible metal dryer hose
Specifications
Type Air Diverter
For Use With (Application) Divert GC Exhaust Heat up and Away From The Lab Bench
Packaging Quantity 1 Each
For Use With Agilent 5890, 6890, 7890 GCs