Learn More
Trajan Scientific and Medical™ Replacement Needles for CTC RTC and Thermo Scientific Plunger-in-Needle Syringes
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
| Description | Replacement Needle |
| Needle Gauge | 23 G |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Volume (Metric) | 0.5 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | CTC RTC and Thermo Scientific Plunger-in-Needle Syringes |