Learn More
BOLA Replacement Inner Parts
Product Code. 10047891
Change view
Click to view available options
Diameter (Metric):
0.8 mm
1.6 mm
3 mm
4 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10047891 0.8 mm Each
10727512 3 mm Each
10430922 4 mm Each
10217171 1.6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10047891 Supplier BOLA Supplier No. D59802
Only null left
Add to basket
Add to basket
Replacement Inner Parts PTFE, PVDF
  • Consisting of a v-ring (ETFE), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm (not exposed to the medium).
  • Very good chemical resistance, the medium is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Diameter (Metric) 0.8 mm
Autoclavable Autoclavable
Thread Size GL 14
Color White
Flexibility Rigid