missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, ACS Reagent

Product Code. 10075640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10075640 100 g Glass bottle 100g
10790292 25 g Glass bottle 25g
10105323 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10075640 Supplier Thermo Scientific Acros Supplier No. 423331000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13573-16-5 | C4H10CrN7S4·H2O | 354.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-16-5
Molecular Formula C4H10CrN7S4·H2O
Molecular Weight (g/mol) 354.46
MDL Number MFCD00150925
Synonym Ammonium reineckate, Ammonium tetrathiocyanatodiamminechromate

Specifications

Melting Point 270.0°C
Color Red
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula NH4(Cr(NH3)2(SCN)4)·H2O
Merck Index 15, 8248
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in deluted hydrochloric acid,soluble in ethanol
Formula Weight 354.46
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Glass bottle
Sensitivity Passes Test
Chemical Name or Material Reinecke salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.