missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus 50S ribosomal protein L7 / L12 Protein

Product Code. 16097900
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16097900 100 μg 100µg
16007910 10 μg 10µg
16047910 50 μg 50µg
16027910 200 μg 200µg
16037910 500 μg 500µg
16017910 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16097900 Supplier enQuireBio™ Supplier No. QP7259ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) 50S ribosomal protein L7/L12, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_001273586.1
Gene ID (Entrez) 31213346
Name S. aureus 50S ribosomal protein L7 / L12 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol rplL
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity S. aureus
Species E. coli
Protein Tag His-SUMO
Sequence MANHEQIIEA IKEMSVLELN DLVKAIEEEF GVTAAAPVAV AGAAGGADAA AEKTEFDVEL TSAGSSKIKV VKAVKEATGL GLKDAKELVD GAPKVIKEAL PKEEAEKLKE QLEEVGATVE LK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.