missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ready-To-Go™ RT-PCR Beads

Product Code. 10064974 Shop All Cytiva Products
Change view
Click to view available options
Format:
0.2mL Tube
0.2mL Tube with hinged cap
0.5mL Tube
No. of Reactions:
96
100
Volume:
200μL
500μL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format No. of Reactions Volume unitSize
10064974 0.2mL Tube 96 200μL Each
10187044 0.2mL Tube with hinged cap 96 200μL Each
10015264 0.5mL Tube 100 500μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10064974 Supplier Cytiva Supplier No. 27926701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimized for performing either one-step or conventional two-step reverse transcription. Cytiva Lifescience™ Illustra™ Ready-To-Go™ RT-PCR Beads are ambient temperature stable, pre-mixed, and ready for pre-dispensed reactions.

  • Reagents are optimized for full-length cDNA synthesis (>7.5 kb) and optimal sensitivity from PCR
  • Provided in either thin walled 0.5 ml or 0.2 ml tubes/plates compatible with most thermocyclers
  • Supplied with both pd(N)6 and oligo(dT) primers for flexibility
  • The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature stable beads containing Buffer, dATP, dCTP, dGTP, dTTP, approximately 2 units of Taq DNA Polymerase, M-MLV reverse Transcriptase, RNAguard (porcine) and stabilizers (including RNase/DNase free BSA), when a bead is reconstituted to 50. Ambient in pouch with desiccant
Format 0.2mL Tube
Quantity 1 Kit
For Use With (Application) RT-PCR
No. of Reactions 96
Volume 200μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.