missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Blotting Paper Filter Discs Grade BF 3

Product Code. 10144421 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10144421 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144421 Supplier Sartorius™ Supplier No. FT3521185
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in he life sciences and diagnostics sectors.

  • Grade BF 3
  • Filter discs
  • Ideal for blotting applications
  • Non-sterile, bulk packed
TRUSTED_SUSTAINABILITY

Specifications

Packaging Type Bulk
Description Filter discs
Diameter (Metric) 185 mm
Color White
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.