missing translation for 'onlineSavingsMsg'
Learn More

rac-1-(2-Nitrophenyl)ethanol, TRC

Product Code. 30479543
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479543 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479543 Supplier Toronto Research Chemicals Supplier No. TRCN5034101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,S)-1-(2-Nitrophenyl)ethanol
CAS 3205-25-2
Molecular Formula C8 H9 N O3
Quantity 1 g
Synonym Benzenemethanol','α-methyl-2-nitro-, Benzyl alcohol','α-methyl-o-nitro- (6CI,7CI,8CI), α-Methyl-2-nitrobenzenemethanol, 1-(2-Nitrophenyl)ethan-1-ol, 1-(2-Nitrophenyl)ethanol, 1-(2-Nitrophenyl)ethyl alcohol, NSC 47195, α-(2-Nitrophenyl)ethanol, α-Methyl-2-nitrobenzyl alcohol, (R,S)-1-(2-Nitrophenyl)ethanol
InChI Formula InChI=1S/C8H9NO3/c1-6(10)7-4-2-3-5-8(7)9(11)12/h2-6,10H,1H3
SMILES CC(O)c1ccccc1[N+](=O)[O-]
IUPAC Name 1-(2-nitrophenyl)ethanol
Molecular Weight (g/mol) 167.16
Formula Weight 167.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.