missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG1 Secondary Antibody (20) [HRP], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43768H
This item is not returnable.
View return policy
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA
Specifications
| IgG1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Mouse | |
| Purified |
| ELISA | |
| 20 | |
| ELISA | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Recombinant Mouse IgG1 Fc Protein | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction