Learn More
Invitrogen™ Rabbit anti-Goat IgG F(ab')2 Secondary Antibody, Biotin
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31753 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31753 reacts with the light chains of goat IgG and with the light chains common to most goat immunoglobulins, but not with the Fc portion of goat IgG or with non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.5 mL of distilled water (1.3 mg/mL after restoration, lot-dependent). Country of Origin: USA.
Specifications
Specifications
| Antigen | Goat IgG F(ab')2 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Rabbit |
| Purification Method | Affinity chromatography |
| Quantity | 1.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.