missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (R)-(-)-Apomorphine hydrochloride
GREENER_CHOICE

Product Code. 30001366
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001366 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001366 Supplier Tocris Bioscience™ Supplier No. 2073/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dopamine agonist; non-subtype-selective

Prototypical dopamine agonist (pKi values are 6.43, 7.08, 7.59, 8.36 and 7.83 for human recombinant D1, D2L, D3, D4 and D5 receptors respectively). Produces biphasic effects on locomotor activity, and displays anti-Parkinsonian and neuroprotective actions following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 314-19-2
Quantity 50 mg
Molecular Formula C17H18ClNO2
Formula Weight 303.79
Purity >99%
Molecular Weight (g/mol) 303.786
Chemical Name or Material (R)-(-)-Apomorphine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.