missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone, 97%

Product Code. 15447957
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447957 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447957 Supplier Thermo Scientific Alfa Aesar Supplier No. H51074.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 173604-33-6 | C16H15NO2 | 253.301 g/mol

(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible w
TRUSTED_SUSTAINABILITY

Specifications

CAS 173604-33-6
Molecular Formula C16H15NO2
MDL Number MFCD01863565
Quantity 1 g
Synonym r-4-benzhydryloxazolidin-2-one, r-+-4-diphenylmethyl-2-oxazolidinone, r-4-methyldiphenyl-2-oxazolidinone, 4r-4-diphenylmethyl-1,3-oxazolidin-2-one, pubchem11686, 4r-4-benzhydryloxazolidine-2-one, r-4-diphenylmethyl oxazolidin-2-one, 2-oxazolidinone,4-diphenylmethyl-, 4r, 2-oxazolidinone, 4-diphenylmethyl-, 4r, r-+-4-diphenylmethyl-2-oxazolidin
Solubility Information Slightly soluble in water.
InChI Key QEOCTJMBYZNEJH-AWEZNQCLSA-N
SMILES C1C(NC(=O)O1)C(C2=CC=CC=C2)C3=CC=CC=C3
IUPAC Name (4R)-4-benzhydryl-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 253.301
PubChem CID 9837924
Formula Weight 253.3
Percent Purity 98%
Chemical Name or Material (R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.