missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Qdot™ 525 Streptavidin Conjugate

Product Code. 10121013
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
50µL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121013 50 μL 50µL
10466673 200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10121013 Supplier Molecular Probes™ Supplier No. Q10143MP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Qdot™ 525 Streptavidin Conjugate

Qdot™ 525 Streptavidin Conjugate comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Qdot™ nanocrystal). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays, and are also used in purification workflows to achieve targeted fractionation. Qdot™ nanocrystal conjugates are provided as 1μM solutions.

Important Features of Qdot™ Streptavidin Conjugates:
  • Has emission maxima of ∼525 nm
  • Approximately 5 to 10 streptavidins per Qdot™ nanocrystal
  • Extremely photostable and bright fluorescence
  • Efficiently excited with single-line excitation sources
  • Narrow emission, large Stokes shift
  • Available in multiple colors
  • Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

    Properties of Qdot™ Nanocrystals
    The Qdot™ streptavidin conjugate is the size of a large macromolecule or protein (∼15–20 nm) and represents the brightest class of streptavidin detection reagents. Qdot™ streptavidin conjugates are made from a nanometer-scale crystal of a semiconductor material (CdSe), which is coated with an additional semiconductor shell (ZnS) to improve the optical properties of the material. The Qdot™ 705 and Qdot™ 800 streptavidin conjugates, which include CdSeTe, are made in a similar fashion. This core-shell material is further coated with a polymer shell that allows the materials to be conjugated to biological molecules and to retain their optical properties.

    Other Fluorescent Conjugates of Streptavidin are Available
    We offer several other Qdot™ colors or try the Qdot™ Streptavidin Sampler Kit, containing Qdot™ streptavidin conjugates in six colors. In addition to nanocrystal conjugates, we offer a wide range of streptavidins conjugated to Alexa Fluor™ dyes, Oregon Green™ dye, enzyme conjugates, and traditional fluorophores like Texas Red™ dye, fluorescein (FITC) and more.

    Find Biotinylated Conjugates
    We offer an extensive line of biotinylated conjugates for use in biotin-streptavidin detection strategies.
  • Use the primary antibody search tool to find biotinylated primary antibodies
  • Use the secondary antibody selector tool to find biotinylated secondary antibodies and biotinylated anti-dye and anti-hapten antibodies

    Blocking Endogenous Biotin
    Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 1 μM
    Conjugate Qdot 525
    Quantity 50 μL
    Content And Storage 1 tube — Store at 2 to 6°C; DO NOT FREEZE
    Shipping Condition Room Temperature
    Form Liquid
    Product Line Qdot
    Product Type Streptavidin Conjugate (fluorescent)

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.