Learn More
Pyrex™ PBT Screwcaps
Product Code. 10034794
Change view
Click to view available options
Thread Size (Metric):
13 mm
15 mm
20 mm
22 mm
25 mm
30 mm
42 mm
Unit Size:
Pack of 10
Pack of 20
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
10034794 25 mm Pack of 20
10300025 13 mm Pack of 20
10165324 15 mm Pack of 20
10399974 20 mm Pack of 20
10389974 22 mm Pack of 20
15985410 30 mm Pack of 20
12579835 30 mm Pack of 20
10181930 42 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10034794 Supplier Pyrex Supplier No. 4503/25
Only null left
Add to basket
Add to basket
Use with reagent bottles, culture tubes and other screwtop bottles. Pyrex™ PBT Screwcaps come in a wide range of sizes for your laboratory needs. Sealing discs provide inert inner seal, resistance to moisture vapor, and are great for culture work.
  • PTFE faced sealing disc for chemical resistance
  • Withstands hot air sterilization to 180deg.C
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description Screw cap
Includes PTFE-faced Rubber Insert
Material PBT
Quantity 20 Pk.
Thread Size (Metric) 25 mm
Liner Material PTFE-faced Rubber
Product Type Screw Cap
For Use With (Equipment) Reagent and media bottles, culture tubes
Autoclavable Autoclavable