missing translation for 'onlineSavingsMsg'
Learn More

Pullulan, MP Biomedicals™

Product Code. 11437392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437392 1 g 1g
11447392 5 g 5g
11457392 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437392 Supplier MP Biomedicals Supplier No. 0215394401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Produced from starch by cultivating a black yeast. Pullulan, MP Biomedicals is an edible plastic with excellent adhesiveness and film formability. It is a natural neutral polysaccharfied used in studies assessing bone tissue engineering and investigating

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9057-02-7
Molecular Formula (C18H30O15)AHO
InChI Key GFZFEWWPMNSVBS-WVZDODFGSA-N
Synonym Pullulan from Aureobasidium pullulans
PubChem CID 131636581
IUPAC Name (2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O

Specifications

Quantity 1 g
Solubility Information Dissolves easily in cold and hot water (50mg/mL; 1/20: Clear).
Formula Weight 200,000 (Lit.)
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.