Learn More
Kartell™ PTFE Supple Connectors
Product Code. 10021061
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
Product Code. Material unitSize
10021061 PTFE Each
1 options
Product Code. 10021061 Supplier Kartell™ Supplier No. 0104800
Only null left
Add to basket
Add to basket
Manufactured from PTFE. Kartell™ PTFE Supple Connectors are specially designed to connect to misaligned glass tubes. The connectors are chemically inert and suitable for food contents.
  • PTFE
  • Suitable for food contents
  • Taper conforms to DIN 12242
  • Chemically inert
Specifications
Certifications/Compliance Yes
Material PTFE
For Use With Misaligned glass tubes
Height (Metric) 29.63 mm
Product Type Supple Connector