Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar
Product Code. 17875294
Change view
Click to view available options
Length (Metric):
6 mm
10 mm
35 mm
Unit Size:
Pack of 10
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
17875294 10 mm Pack of 20
10490194 10 mm Pack of 10
10637952 35 mm Pack of 10
10057661 6 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17875294 Supplier Bohlender™ Supplier No. C35008
Only null left
Add to basket
Add to basket
Specifications
Certifications/Compliance FDA Certified
For Use With (Application) Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
Includes Alnico 5 magnetic core
Diameter (Metric) 3 mm
Length (Metric) 10 mm
Material PTFE
Quantity 1/Pk.
Shape Cylinder