missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™

Product Code. 10166924 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10166924 100 mL AcroSeal™ Glass Bottle 100mL
15179293 1 L AcroSeal™ Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10166924 Supplier Thermo Scientific Acros Supplier No. 429121000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-96-7 | C3H3Br | 118.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 88.0°C to 90.0°C
Chemical Name or Material Propargyl bromide
Name Note Stabilized
CAS 106-96-7, 108-88-3
Color White to Yellow
Physical Form Crystalline Powder
Quantity 100 mL
Molecular Formula C3H3Br
Linear Formula HC≡CCH2Br
MDL Number MFCD00000241
Flash Point 4°C
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
Solubility Information Solubility in water: immiscible
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
SMILES C#CCBr
IUPAC Name 3-bromoprop-1-yne
Molecular Weight (g/mol) 118.96
PubChem CID 7842
Formula Weight 118.96
Percent Purity 73 to 87% (propargyl bromide) (GC), 13 to 27% (toluene) (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4900 to 1.4960
Specific Gravity 1.38
Density 1.3800g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl bromide, 80 wt% solution in toluene, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.