missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Universal Assay Buffer (10X)
Click to view available options
Quantity:
1 each
Unit Size:
Each
Specifications
Specifications
| Product Type | Buffer |
| Quantity | 1 each |
| Species | Canine, Mouse, Rat, Non-Human Primate, Human, Human |
| For Use With (Equipment) | Luminex™ Instruments |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction