missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRIMER,5'M13 50 NMOL
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| For Use With (Equipment) | Normal or Reversed-phase Chromatography, HPLC or PAGE |
| Description | Custom 5'M13 Primer, 50 nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification |
| Product Type | Custom 5'M13 Primer |
| Quantity | 50 nmol |
| Grade | HPLC |
| For Use With (Application) | Design a Custom DNA Oligonucleotide, with any of a variety of 3' and 5' Modification |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?