missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRICKLE1 Polyclonal antibody specifically detects PRICKLE1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | PRICKLE1 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EPM1BMGC138902, FLJ31627, FLJ31937, prickle homolog 1 (Drosophila), prickle-like 1, prickle-like 1 (Drosophila), prickle-like protein 1, REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencerfactor)-interacting LIM domain protein, REST/NRSF-interacting LIM domain protein 1, RILPMGC138903 |
| Host Species | Rabbit |
| Immunogen | prickle homolog 1(Drosophila). (Uniprot# Q96MT3) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?