missing translation for 'onlineSavingsMsg'
Learn More

PPWD1 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30763456 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763456 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763456 Supplier Novus Biologicals Supplier No. NBP346905MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPWD1 Polyclonal antibody specifically detects PPWD1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPWD1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias EC 5.2.1.8, KIAA0073Spliceosome-associated cyclophilin, peptidylprolyl isomerase domain and WD repeat containing 1, peptidylprolyl isomerase domain and WD repeat-containing protein 1
Host Species Rabbit
Immunogen peptidylprolyl isomerase domain and WD repeat containing 1. (Uniprot# Q96BP3)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23398
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.